3-Acetyl-5-bromo-1H-indazole 96%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(5-Bromo-1H-indazol-3-yl)ethanone, 1-(5-Bromo-1H-indazol-3-yl)ethan-1-one, 3-Ethanoyl-5-bromo-1H-indazole
OR305110-1G OR305110-250MG
APOSOR305110-1GEA 87 EUR
APOSOR305110-1G APOSOR305110-250MG
3-Acetyl-5-bromo-1H-indazole 96%
3-Acetyl-5-bromo-1H-indazole
Formula: C₉H₇BrN₂O
MW: 239.069 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07374362
CAS Number: 886363-74-2