7-Iodoimidazo[1,2-a]pyridine 95%
Supplier: Apollo Scientific
OR305166-1G OR305166-250MG
APOSOR305166-1GEA 229 EUR
APOSOR305166-1G APOSOR305166-250MG
7-Iodoimidazo[1,2-a]pyridine 95%
7-Iodoimidazo[1,2-a]pyridine
Formula: C₇H₅IN₂
MW: 244.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13177252
CAS Number: 908269-30-7