5-Amino-3H-imidazo[4,5-b]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 3H-Imidazo[4,5-b]pyridin-5-amine, 6-Amino-7-azabenzimidazole
OR305186-250MG OR305186-100MG
APOSOR305186-250MGEA 244 EUR
APOSOR305186-250MG APOSOR305186-100MG
5-Amino-3H-imidazo[4,5-b]pyridine
5-Amino-3H-imidazo[4,5-b]pyridine
Formula: C₆H₆N₄
MW: 134.139 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD11846337
CAS Number: 69825-84-9