1H-Indole-4-carbohydrazide
Supplier: Apollo Scientific
OR305209-1G OR305209-250MG
APOSOR305209-1GEA 200 EUR
APOSOR305209-1G APOSOR305209-250MG
1H-Indole-4-carbohydrazide
1H-Indole-4-carbohydrazide
Formula: C₉H₉N₃O
MW: 175.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738885
CAS Number: 885272-22-0