7-Chloro-5-methyl-1H-pyrazolo[4,3-b]pyridine
Supplier: Apollo Scientific
Synonyms: 7-Chloro-5-methyl-1H-pyrazolo[4,3-b]-pyridine
OR305333-250MG OR305333-100MG OR305333-1G
APOSOR305333-250MGEA 197 EUR
APOSOR305333-250MG APOSOR305333-100MG APOSOR305333-1G
7-Chloro-5-methyl-1H-pyrazolo[4,3-b]pyridine
7-Chloro-5-methyl-1H-pyrazolo[4,3-b]pyridine
Formula: C₇H₆ClN₃
MW: 167.596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08236755
CAS Number: 94220-38-9