1H-Pyrazolo[4,3-c]pyridine-6-carboxylic acid 97%
Supplier: Apollo Scientific
OR305341-250MG OR305341-1G
APOSOR305341-250MGEA 453 EUR
APOSOR305341-250MG APOSOR305341-1G
1H-Pyrazolo[4,3-c]pyridine-6-carboxylic acid 97%
1H-Pyrazolo[4,3-c]pyridine-6-carboxylic acid
Formula: C₇H₅N₃O₂
MW: 163.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18260541
CAS Number: 1256802-03-5