5-Phenyl-1,3-thiazol-2-amine
Supplier: Apollo Scientific
Synonyms: 5-Phenylthiazol-2-amine, 2-Amino-5-phenylthiazole, 5-Phenyl-1,3-thiazol-2-amine, 5-Phenyl-2-thiazolamine
OR305428-1G OR305428-250MG
APOSOR305428-1GEA 101 EUR
APOSOR305428-1G APOSOR305428-250MG
5-Phenyl-1,3-thiazol-2-amine
5-Phenyl-1,3-thiazol-2-amine
Formula: C₉H₈N₂S
MW: 176.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022451
CAS Number: 39136-63-5