Methyl 8-oxo-1,4-dioxaspiro[4.5]decane-7-carboxylate 96%
Supplier: Apollo Scientific
Warning
OR305438-1G OR305438-250MG
APOSOR305438-1GEA 284 EUR
APOSOR305438-1G APOSOR305438-250MG
Methyl 8-oxo-1,4-dioxaspiro[4.5]decane-7-carboxylate 96%
Methyl 8-oxo-1,4-dioxaspiro[4.5]decane-7-carboxylate
Formula: C₁₀H₁₄O₅
MW: 214.215 g/mol
Melting Pt: 60…61 °C
Storage Temperature: Ambient
MDL Number: MFCD03271523
CAS Number: 52506-21-5