4-(Propan-2-yl)-1H-pyrazol-3-amine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Isopropyl-1H-pyrazol-5-amine
OR305561-1G OR305561-250MG OR305561-5G
APOSOR305561-1GEA 133 EUR
APOSOR305561-1G APOSOR305561-250MG APOSOR305561-5G
4-(Propan-2-yl)-1H-pyrazol-3-amine 95%
4-(Propan-2-yl)-1H-pyrazol-3-amine
Formula: C₆H₁₁N₃
MW: 125.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112127
CAS Number: 151521-49-2