4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Supplier: Apollo Scientific
Warning
OR30658-1G OR30658-250MG
APOSOR30658-1GEA 94.4 EUR
APOSOR30658-1G APOSOR30658-250MG
4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
4,6-Dichloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₆H₄Cl₂N₄
MW: 203.029 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09750213
CAS Number: 98141-42-5