4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine
Supplier: Apollo Scientific
Warning
OR30659-250MG OR30659-1G
APOSOR30659-250MGEA 198 EUR
APOSOR30659-250MG APOSOR30659-1G
4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine
4,6-Dichloro-1,3-dimethyl-1H-pyrazolo[3,4-d]pyrimidine
Formula: C₇H₆Cl₂N₄
MW: 217.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11869749
CAS Number: 1072895-86-3