N-Boc-(R)-2-Amino-3-methoxy-1-propanol 95%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl [(2R)-1-hydroxy-3-methoxypropan-2-yl]carbamate, (R)-tert-Butyl (1-hydroxy-3-methoxypropan-2-yl)carbamate
OR307123-1G OR307123-5G OR307123-25G
APOSOR307123-1GEA 33.5 EUR
APOSOR307123-1G APOSOR307123-5G APOSOR307123-25G
N-Boc-(R)-2-Amino-3-methoxy-1-propanol 95%
N-Boc-(R)-2-Amino-3-methoxy-1-propanol
Formula: C₉H₁₉NO₄
MW: 205.251 g/mol
Boiling Pt: 95…97 °C (0.8 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD20261174
CAS Number: 183793-49-9