Ethyl 3-iodo-1H-pyrazole-4-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 4-(Ethoxycarbonyl)-3-iodo-1H-pyrazole
OR307164-1G OR307164-250MG OR307164-5G
APOSOR307164-1GEA 66.7 EUR
APOSOR307164-1G APOSOR307164-250MG APOSOR307164-5G
Ethyl 3-iodo-1H-pyrazole-4-carboxylate
Ethyl 3-iodo-1H-pyrazole-4-carboxylate
Formula: C₆H₇IN₂O₂
MW: 266.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072591
CAS Number: 827316-43-8