1H-Pyrazole-4-carbaldehyde hydrochloride
Supplier: Apollo Scientific
Warning
OR307174-1G OR307174-5G
APOSOR307174-1GEA 50.9 EUR
APOSOR307174-1G APOSOR307174-5G
1H-Pyrazole-4-carbaldehyde hydrochloride
1H-Pyrazole-4-carbaldehyde hydrochloride
Formula: C₄H₄N₂O·ClH
MW: 132.548 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09759205
CAS Number: 1197230-88-8