trans-2-Aminomethyl-cyclohexylamine
Supplier: Apollo Scientific
Danger
OR307559-1G OR307559-250MG
APOSOR307559-1GEA 1190 EUR
APOSOR307559-1G APOSOR307559-250MG
trans-2-Aminomethyl-cyclohexylamine
trans-2-Aminomethyl-cyclohexylamine
Formula: C₇H₁₆N₂
MW: 128.215 g/mol
Boiling Pt: 87…88 °C
Storage Temperature: Ambient
MDL Number: MFCD06799044
CAS Number: 24716-89-0
UN: 2735
ADR: 8,II