1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
OR307564-1G OR307564-250MG
APOSOR307564-1GEA 525 EUR
APOSOR307564-1G APOSOR307564-250MG
1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
1-Carboxymethyl-6,7-dimethoxy-3,4-dihydro-1H-isoquinoline-2-carboxylic acid tert-butyl ester
Formula: C₁₈H₂₅NO₆
MW: 351.394 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258126
CAS Number: 282524-92-9