cis-2-tert-Butoxycarbonylamino-trans-5-(4-chlorophenyl)-cyclohexanecarboxylic acid
Supplier: Apollo Scientific
OR307565-1G OR307565-250MG
APOSOR307565-1GEA 781 EUR
APOSOR307565-1G APOSOR307565-250MG
cis-2-tert-Butoxycarbonylamino-trans-5-(4-chlorophenyl)-cyclohexanecarboxylic acid
cis-2-tert-Butoxycarbonylamino-trans-5-(4-chlorophenyl)-cyclohexanecarboxylic acid
Formula: C₁₈H₂₄ClNO₄
MW: 353.84 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863629
CAS Number: 1176474-84-2