cis-2-tert-Butoxycarbonylamino-2-methyl-cyclohexanecarboxylic acid
Supplier: Apollo Scientific
OR307583-1G OR307583-250MG
APOSOR307583-1GEA 507 EUR
APOSOR307583-1G APOSOR307583-250MG
cis-2-tert-Butoxycarbonylamino-2-methyl-cyclohexanecarboxylic acid
cis-2-tert-Butoxycarbonylamino-2-methyl-cyclohexanecarboxylic acid
Formula: C₁₃H₂₃NO₄
MW: 257.326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863638
CAS Number: 1175673-63-8