cis-(2-Amino-2-methyl-cyclopentyl)-methanol hydrochloride
Supplier: Apollo Scientific
Warning
OR307598-1G OR307598-250MG
APOSOR307598-1GEA 897 EUR
APOSOR307598-1G APOSOR307598-250MG
cis-(2-Amino-2-methyl-cyclopentyl)-methanol hydrochloride
cis-(2-Amino-2-methyl-cyclopentyl)-methanol hydrochloride
Formula: C₇H₁₅NO·ClH
MW: 165.661 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783056
CAS Number: 1212406-48-8