cis-2-Aminomethyl-1-methyl-cyclohexanol hydrochloride
Supplier: Apollo Scientific
OR307599-1G OR307599-250MG
APOSOR307599-1GEA 590 EUR
APOSOR307599-1G APOSOR307599-250MG
cis-2-Aminomethyl-1-methyl-cyclohexanol hydrochloride
cis-2-Aminomethyl-1-methyl-cyclohexanol hydrochloride
Formula: C₈H₁₇NO·ClH
MW: 179.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07783752
CAS Number: 1212419-83-4