6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester
Supplier: Apollo Scientific
OR307616-1G OR307616-250MG
APOSOR307616-1GEA 739 EUR
APOSOR307616-1G APOSOR307616-250MG
6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester
6,7-Dimethoxy-3,4-1H-isoquinoline-1-dicarboxylic acid 2-tert-butyl ester
Formula: C₁₇H₂₃NO₆
MW: 337.368 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863624
CAS Number: 738629-59-9