(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-3-yl)-methanol hydrochloride
Supplier: Apollo Scientific
OR307631-1G OR307631-250MG
APOSOR307631-1GEA 1100 EUR
APOSOR307631-1G APOSOR307631-250MG
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-3-yl)-methanol hydrochloride
(6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-3-yl)-methanol hydrochloride
Formula: C₁₂H₁₇NO₃·ClH
MW: 259.729 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11505885
CAS Number: 886997-74-6