(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propan-1-ol
Supplier: Apollo Scientific
OR307633-1G OR307633-250MG
APOSOR307633-1GEA 590 EUR
APOSOR307633-1G APOSOR307633-250MG
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propan-1-ol
(R*)-2-((R*)-6,7-Dimethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-propan-1-ol
Formula: C₁₄H₂₁NO₃
MW: 251.321 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703290
CAS Number: 142976-45-2