cis-(2-Hydroxymethyl-cyclohexyl)-carbamic acid tert-butyl ester
Supplier: Apollo Scientific
OR307680-1G OR307680-250MG
APOSOR307680-1GEA 540 EUR
APOSOR307680-1G APOSOR307680-250MG
cis-(2-Hydroxymethyl-cyclohexyl)-carbamic acid tert-butyl ester
cis-(2-Hydroxymethyl-cyclohexyl)-carbamic acid tert-butyl ester
Formula: C₁₂H₂₃NO₃
MW: 229.316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04974362
CAS Number: 179686-77-2