trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Supplier: Apollo Scientific
OR307700-1G OR307700-250MG
APOSOR307700-1GEA 509 EUR
APOSOR307700-1G APOSOR307700-250MG
trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
trans-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Formula: C₁₃H₁₆ClNO₂
MW: 253.725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368284
CAS Number: 1416307-75-9