cis-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Supplier: Apollo Scientific
OR307701-1G OR307701-250MG
APOSOR307701-1GEA 509 EUR
APOSOR307701-1G APOSOR307701-250MG
cis-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
cis-tert-Butyl 3-(4-chlorophenyl)aziridine-2-carboxylate
Formula: C₁₃H₁₆ClNO₂
MW: 253.725 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368285
CAS Number: 1431364-55-4