7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester
Supplier: Apollo Scientific
OR307812-1G OR307812-5G
APOSOR307812-1GEA 235 EUR
APOSOR307812-1G APOSOR307812-5G
7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester
7-Chloro-2-methyl-quinoline-3-carboxylic acid ethyl ester
Formula: C₁₃H₁₂ClNO₂
MW: 249.693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863550
CAS Number: 733719-74-9