6-Chloro-4-hydroxy-8-methylquinoline
Supplier: Apollo Scientific
OR307829-1G OR307829-250MG
APOSOR307829-1GEA 261 EUR
APOSOR307829-1G APOSOR307829-250MG
6-Chloro-4-hydroxy-8-methylquinoline
6-Chloro-4-hydroxy-8-methylquinoline
Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272439
CAS Number: 203626-38-4