2-Chloro-3-phenylquinoline
Supplier: Apollo Scientific
OR307834-1G OR307834-5G
APOSOR307834-1GEA 293 EUR
APOSOR307834-1G APOSOR307834-5G
2-Chloro-3-phenylquinoline
2-Chloro-3-phenylquinoline
Formula: C₁₅H₁₀ClN
MW: 239.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179790
CAS Number: 2859-30-5