2-Chloro-5,7-dimethylquinoline-3-carboxaldehyde
Supplier: Apollo Scientific
OR307912-1G OR307912-5G
APOSOR307912-1GEA 259 EUR
APOSOR307912-1G APOSOR307912-5G
2-Chloro-5,7-dimethylquinoline-3-carboxaldehyde
2-Chloro-5,7-dimethylquinoline-3-carboxaldehyde
Formula: C₁₂H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD02227047
CAS Number: 482639-32-7