4-Hydrazino-6-methoxyquinoline hydrochloride
Supplier: Apollo Scientific
OR307975-1G OR307975-5G
APOSOR307975-1GEA 301 EUR
APOSOR307975-1G APOSOR307975-5G
4-Hydrazino-6-methoxyquinoline hydrochloride
4-Hydrazino-6-methoxyquinoline hydrochloride
Formula: C₁₀H₁₁N₃O·ClH
MW: 225.675 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09787749
CAS Number: 68500-39-0