4-Hydroxy-6-methoxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR307977-1G OR307977-5G
APOSOR307977-1GEA 60.9 EUR
APOSOR307977-1G APOSOR307977-5G
4-Hydroxy-6-methoxyquinoline-3-carboxylic acid
4-Hydroxy-6-methoxyquinoline-3-carboxylic acid
Formula: C₁₁H₉NO₄
MW: 219.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024003
CAS Number: 28027-16-9