4-Hydroxy-8-methylquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR307981-1G OR307981-5G
APOSOR307981-1GEA 194 EUR
APOSOR307981-1G APOSOR307981-5G
4-Hydroxy-8-methylquinoline-3-carboxylic acid
4-Hydroxy-8-methylquinoline-3-carboxylic acid
Formula: C₁₁H₉NO₃
MW: 203.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01909986
CAS Number: 35966-17-7