5,7-Dimethyl-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR307987-1G OR307987-5G
APOSOR307987-1GEA 293 EUR
APOSOR307987-1G APOSOR307987-5G
5,7-Dimethyl-4-hydroxyquinoline-3-carboxylic acid
5,7-Dimethyl-4-hydroxyquinoline-3-carboxylic acid
Formula: C₁₂H₁₁NO₃
MW: 217.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07644537
CAS Number: 948293-86-5