6,8-Dichloro-4-hydroxy-2-methylquinoline
Supplier: Apollo Scientific
OR307999-1G OR307999-5G
APOSOR307999-1GEA 293 EUR
APOSOR307999-1G APOSOR307999-5G
6,8-Dichloro-4-hydroxy-2-methylquinoline
6,8-Dichloro-4-hydroxy-2-methylquinoline
Formula: C₁₀H₇Cl₂NO
MW: 228.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100263
CAS Number: 95541-31-4