(2R)-2-(Aminomethyl)piperidine, N1-BOC protected
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl (2R)-2-(aminomethyl)piperidine-1-carboxylate
OR308131-1G OR308131-5G
APOSOR308131-1GEA 301 EUR
APOSOR308131-1G APOSOR308131-5G
(2R)-2-(Aminomethyl)piperidine, N1-BOC protected
(2R)-2-(Aminomethyl)piperidine, N1-BOC protected
Formula: C₁₁H₂₂N₂O₂
MW: 214.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659040
CAS Number: 683233-14-9