(3R)-3-(Propan-2-yl)morpholine
Supplier: Apollo Scientific
Synonyms: (R)-3-Isopropylmorpholine
OR308181-1G OR308181-250MG
APOSOR308181-1GEA 249 EUR
APOSOR308181-1G APOSOR308181-250MG
(3R)-3-(Propan-2-yl)morpholine
(3R)-3-(Propan-2-yl)morpholine
Formula: C₇H₁₅NO
MW: 129.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519111
CAS Number: 74572-01-3