5-Hydroxy-2-methyl-2H-indazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methyl-2H-indazol-5-ol
OR30847-100MG OR30847-250MG OR30847-1G
APOSOR30847-100MGEA 192 EUR
APOSOR30847-100MG APOSOR30847-250MG APOSOR30847-1G
5-Hydroxy-2-methyl-2H-indazole
5-Hydroxy-2-methyl-2H-indazole
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028572
CAS Number: 1159511-41-7