4-(Hydroxymethyl)-1H-indazole 97%
Supplier: Apollo Scientific
Warning
Synonyms: (1H-Indazol-4-yl)methanol
OR30858-1G OR30858-5G
APOSOR30858-1GEA 26.1 EUR
APOSOR30858-1G APOSOR30858-5G
4-(Hydroxymethyl)-1H-indazole 97%
4-(Hydroxymethyl)-1H-indazole
Formula: C₈H₈N₂O
MW: 148.162 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD11052642
CAS Number: 709608-85-5