4,8-Dichloro-2-trichloromethylquinoline
Supplier: Apollo Scientific
OR309242-1G OR309242-5G
APOSOR309242-1GEA 285 EUR
APOSOR309242-1G APOSOR309242-5G
4,8-Dichloro-2-trichloromethylquinoline
4,8-Dichloro-2-trichloromethylquinoline
Formula: C₁₀H₄Cl₅N
MW: 315.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272425
CAS Number: 93600-66-9