2-Chloro-7-methoxy-3-phenylquinoline
Supplier: Apollo Scientific
OR309264-1G OR309264-5G
APOSOR309264-1GEA 293 EUR
APOSOR309264-1G APOSOR309264-5G
2-Chloro-7-methoxy-3-phenylquinoline
2-Chloro-7-methoxy-3-phenylquinoline
Formula: C₁₆H₁₂ClNO
MW: 269.726 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998022
CAS Number: 85274-56-2