6-Chloro-4-hydroxy-2-methylquinoline
Supplier: Apollo Scientific
Synonyms: 6-Chloro-2-methylquinolin-4-ol
OR309278-1G OR309278-5G
APOSOR309278-1GEA 74.1 EUR
APOSOR309278-1G APOSOR309278-5G
6-Chloro-4-hydroxy-2-methylquinoline
6-Chloro-4-hydroxy-2-methylquinoline
Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023981
CAS Number: 15644-86-7