8-Chloro-4-hydroxy-2-phenylquinoline
Supplier: Apollo Scientific
OR309301-1G OR309301-5G
APOSOR309301-1GEA 301 EUR
APOSOR309301-1G APOSOR309301-5G
8-Chloro-4-hydroxy-2-phenylquinoline
8-Chloro-4-hydroxy-2-phenylquinoline
Formula: C₁₅H₁₀ClNO
MW: 255.699 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02929094
CAS Number: 439147-12-3