tert-Butyl 4-{2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate
Supplier: Apollo Scientific
OR310061-1G OR310061-500MG
APOSOR310061-1GEA 538 EUR
APOSOR310061-1G APOSOR310061-500MG
tert-Butyl 4-{2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate
tert-Butyl 4-{2-oxo-1H,2H,3H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate
Formula: C₁₆H₂₂N₄O₃
MW: 318.371 g/mol
Melting Pt: 201…204 °C
Storage Temperature: Ambient
MDL Number: MFCD13184781
CAS Number: 781649-87-4