2-Chloro-5-methyl-4-(methylthio)pyrimidine
Supplier: Apollo Scientific
Warning
OR31018-1G OR31018-250MG OR31018-5G
APOSOR31018-1GEA 132 EUR
APOSOR31018-1G APOSOR31018-250MG APOSOR31018-5G
2-Chloro-5-methyl-4-(methylthio)pyrimidine
2-Chloro-5-methyl-4-(methylthio)pyrimidine
Formula: C₆H₇ClN₂S
MW: 174.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21790275
CAS Number: 213334-25-9