4-Chlorothieno[3,2-d]pyrimidine 97%
Supplier: Apollo Scientific
Warning
OR3102-25G OR3102-500MG
APOSOR3102-25GEA 134 EUR
APOSOR3102-25G APOSOR3102-500MG
4-Chlorothieno[3,2-d]pyrimidine 97%
4-Chlorothieno[3,2-d]pyrimidine
Formula: C₆H₃ClN₂S
MW: 170.619 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00122144
CAS Number: 16269-66-2