9-Methyl-3,9-diazabicyclo[4.2.1]nonan-4-one
Supplier: Apollo Scientific
Warning
OR310326-1G OR310326-500MG
APOSOR310326-1GEA 1070 EUR
APOSOR310326-1G APOSOR310326-500MG
9-Methyl-3,9-diazabicyclo[4.2.1]nonan-4-one
9-Methyl-3,9-diazabicyclo[4.2.1]nonan-4-one
Formula: C₈H₁₄N₂O
MW: 154.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08693835
CAS Number: 7309-42-4