8-Phenyl-1,4-dioxaspiro[4.5]decane-8-carbaldehyde
Supplier: Apollo Scientific
OR310341-1G OR310341-500MG
APOSOR310341-1GEA 894 EUR
APOSOR310341-1G APOSOR310341-500MG
8-Phenyl-1,4-dioxaspiro[4.5]decane-8-carbaldehyde
8-Phenyl-1,4-dioxaspiro[4.5]decane-8-carbaldehyde
Formula: C₁₅H₁₈O₃
MW: 246.302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26399860
CAS Number: 56327-24-3