1-(Prop-2-en-1-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Supplier: Apollo Scientific
Synonyms: 1-Allyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
OR310846-1G OR310846-5G
APOSOR310846-1GEA 112 EUR
APOSOR310846-1G APOSOR310846-5G
1-(Prop-2-en-1-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-(Prop-2-en-1-yl)-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula: C₁₂H₁₉BN₂O₂
MW: 234.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659011
CAS Number: 1000801-78-4