5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
Supplier: Apollo Scientific
OR310870-1G OR310870-5G
APOSOR310870-1GEA 27.7 EUR
APOSOR310870-1G APOSOR310870-5G
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
5,5-Dimethyl-2-(3-methylphenyl)-1,3,2-dioxaborinane
Formula: C₁₂H₁₇BO₂
MW: 204.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28101508
CAS Number: 223799-24-4